Principaux documents
Z380350
Slant rack for culture tubes
Holds 40 x culture tubes
Synonyme(s) :
rack test tube, racks tube, test tube racks
1 EA
212,00 $
4 EA
625,00 $
212,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
212,00 $
4 EA
625,00 $
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.22
212,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
Holds 40 x culture tubes
Conditionnement
pack of 1 ea
case of 4 ea
Fabricant/nom de marque
Nalgene® 5935-0020
Description générale
Unique end plate design allows 5° or 20° slant when rack is set down on side. Autoclave media upright, then allow to cool on a side. Media will harden with perfect, consistent slant. Also useful for incubating liquids on a slant for better aeration.
Rack holds 40 tubes (4 × 10 array); keyed with numbers and letters. Stackable; will not float in water bath. Autoclaving will reduce strength over time.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Les clients ont également consulté
Slide 1 of 2
1 of 2