Principaux documents
Z360457
Utility boxes, plastic
L × W × H 129 mm × 75 mm × 59 mm
Synonyme(s) :
laboratory storage, plastic storage box
About This Item
Code UNSPSC :
41123300
Nomenclature NACRES :
NB.51
Matériaux
styrene-acrylonitrile (SAN)
Fabricant/nom de marque
Nalgene 5700-0325
L × l × H
129 mm × 75 mm × 59 mm
Description générale
Practical containers for multipurpose lab storage. Glass-clear SAN body; white LDPE lid with white SAN insert. Ideal for dry materials like powders, chromatography sheets. Rounded corners for easy cleaning; stackable for compact freezer storage. Lids snap into place, but are not leakproof.
Dimensions are L × W × H