Principaux documents
Z352543
Sigma-Aldrich
Glass plates for Sigma-Aldrich® sequencing units
size 20 cm × 42 cm
About This Item
Code UNSPSC :
41105600
Nomenclature NACRES :
NB.22
Taille
20 cm × 42 cm
Caractéristiques et avantages
Sets include one rectangular and one notched plate.
Informations légales
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany