Principaux documents
Z329932
BRAND® narrow-mouth bottle, LDPE
capacity 20 mL
Synonyme(s) :
ldpe bottle, polyethylene bottles, polyethylene containers
About This Item
Code UNSPSC :
41121800
Matériaux
low-density polyethylene bottle
narrow-mouth bottle
round bottle
translucent
Description
dropper nozzle caps available
Caractéristiques
cap
Conditionnement
pkg of 100
Fabricant/nom de marque
BRAND 129416
Capacité
20 mL
Diam. × H
31 mm × 58 mm
Raccords
GL 14 thread
Description générale
Sparsame Vielzweck Flasche
Autres remarques
BR129416 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG