Principaux documents
TMO8000-0140
Nalgene® 180 vacuum tubing
tube I.D. × O.D. 3/8 in. × 5/8 in., tube size 50 ft
Synonyme(s) :
plastic vacuum tubing
About This Item
Code UNSPSC :
41121800
Fabricant/nom de marque
Nalge-Nunc 8000-0140
Tube, D.I. × D.E.
3/8 in. × 5/8 in.
Taille du tube
50 ft
Taille de la paroi
1/8 in.
Description générale
Transparent, nontoxic, autoclavable plastic tubing has thick walls for high vacuum.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries