Principaux documents
TMO382019-2000
Nalgene® Certified Clean bottles
clear PETG, volume 2000 mL, case of 12 ea
Synonyme(s) :
PETG bottle, certified nalgene bottle
12 EACH
656,00 $
656,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
12 EACH
656,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
656,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
clear PETG
Conditionnement
case of 12 ea
Fabricant/nom de marque
Nalgene 382019-2000
Capacité
2000 mL
Volume
2000 mL
Description générale
Protect against leakage, breakage and radiation contamination in cleanroom applications with Thermo Scientific Nalgene PETG Certified Clean Containers. Suitable for pharmaceutical and biotechnology applications that require processing and storing bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations. A safe alternative for your lab with no sacrifice of accuracy.
Caractéristiques et avantages
  • Lot-certified for low particulates, these containers features a unique leakproof† design and are suitable for transport.
  • Exhibit excellent gas barrier properties and ensure pH stability for buffered solutions.
  • Resins are free of animal-derived components.
  • Sterile, tamper-resistant seal ensures product integrity prior to use.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries