Principaux documents
TMO362089-0008
Nalgene® narrow-mouth bottle without cap
natural high-density polyethylene, volume 250 mL, case of 250 ea
Synonyme(s) :
Nalgene bottle, plastic bottle
250 EACH
378,00 $
378,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
250 EACH
378,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
378,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
natural high-density polyethylene
Conditionnement
case of 250 ea
Fabricant/nom de marque
Nalgene 362089-0008
Capacité
250 mL
Volume
250 mL
Description générale
Safely store and handle test kits and many small volume applications with Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries