Key Documents
TMO362006-9025
Nalgene® narrow-mouth bottle without cap
amber polypropylene copolymer, volume 8 mL, case of 2000 ea
Synonyme(s) :
Nalgene bottle, plastic bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
amber polypropylene copolymer
Conditionnement
case of 2000 ea
Fabricant/nom de marque
Nalgene 362006-9025
Capacité
8 mL
Volume
8 mL
Description générale
Safely store and handle test kits and many small volume applications with Thermo Scientific Nalgene Natural PPCO Diagnostic Bottles, for use in sample storage and the shipping of small volume, valuable reagents. These polypropylene copolymer bottles are durable and feature uniform walls resistant to splitting or puncturing. They are packed without closures so they can be matched with various colored closures (Cat. No. 362150 series).
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries