Documents
MTCC2086
MTC Bio Stop-Pops Microcentrifuge tube Locking Clips
for 1.5 mL tubes
Synonyme(s) :
centrifuge tube clips, tube closures
About This Item
Code UNSPSC :
41122500
Nomenclature NACRES :
NB.78
Matériaux
polypropylene
Conditionnement
pack of 100 ea
Fabricant/nom de marque
MTC Bio C2086
Capacité
1.5 mL
Description générale
Stop-Pops Locking Clips, w/ breakaway lifting tabs for 1.5mL tubes, 100/pk
Prevent microtubes from popping open during use with this secure locking clip.
Informations légales
MTC is a trademark of MTC Bio
Stop-Pops is a trademark of MTC Bio