Principaux documents
GN384070
Greiner CapMat for sealing multiwell plates
384 well, square nap, silicone rubber, translucent
Synonyme(s) :
Greiner CapMats for sealing multiwell plates
About This Item
Code UNSPSC :
41122410
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Greiner 384070
Disponibilité
available only in China
Résistance chim./phys.
Greiner CapMats are DMSO-resistant and withstand temperatures from -20°C to +60°C