Principaux documents
GE27-5340-01
AutoSeq G-50
Cytiva 27-5340-01, pack of 50 columns
1 PKG
286,00 $
286,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
286,00 $
About This Item
Code UNSPSC :
41105501
Nomenclature NACRES :
NA.52
286,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 50 columns
Fabricant/nom de marque
Cytiva 27-5340-01
Taille des particules
50-150 μm (dry)
Description générale
AutoSeq G-50 columns are specifically designed to remove fluorescent dye-terminators from cycle sequencing reactions prior to analysis on automated sequencers. Effective DNA purification is essential for high-quality results on automated DNA sequencing since residual dye-terminators can obscure data. AutoSeq G-50 consists of MicroSpin columns containing Sephadex G-50 DNA Grade F, pre-equilibrated in double-distilled water. This is important because even the small amount of salt in traditional buffers (such as TE buffer) can cause electrophoretic artefacts on salt-sensitive automated DNA sequencers.
Caractéristiques et avantages
  • For rapid removal of unincorporated fluorescent dye-terminators from cycle sequencing reactions prior to automated DNA sequencing
  • Columns are prepacked with Sephadex G-50 DNA Grade F
  • Columns are pre-equilibrated in double-distilled water with 0.05% Kathon CG/ICP Biocide added as a preservative
  • Ready-to-use columns enable sample application through to collection of purified product in less than 4 min
  • 4 minutes
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Remarque sur l'analyse
To view the Certificate of Analysis for this product, please visit www.cytiva.com.
Informations légales
AutoSeq is a trademark of Cytiva