Principaux documents
GE25-9005-98
Autoscreen A, 96-Well Plate Pack
About This Item
Code UNSPSC :
41106305
Nomenclature NACRES :
NA.52
Le produit GE25-9005-98 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Fabricant/nom de marque
Cytiva
Conditionnement
pkg of (10 x 96-well plates)
Description générale
AutoScreen 96-Well Plate consists of a 96-well filter plate containing DNA Grade Sephadex G-50 for purification of sequencing reactions prior to analysis on ABI sequencers and can be used for other size exclusion applications.
Stockage et stabilité
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.