Principaux documents
GBL204974
Grace Bio-Labs FlexWell SealStrips
24 mm x 77 mm seal area, non-sterile
Synonyme(s) :
polyolefin slide cover, slide seal
50 EA
92,70 $
92,70 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
50 EA
92,70 $
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.13
92,70 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polyolefin
Stérilité
non-sterile
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Grace Bio-Labs 204974
Taille externe
24 mm × 77 mm
Description générale
Transparent, non-fluorescing polyolefin film coated with a hydrophobic adhesive: Non-fluorescing feature will not interfere with automated analyses with fluorometers; and provided on a white paper release liner. Non-sterile,  working surface RNase/DNase –Free
Autres remarques
SealStrip applicator (non-pyrogenic , polyethylene) included (one per pack of 50)
Informations légales
FlexWell is a trademark of Grace Bio-Labs, Inc.
SealStrips is a trademark of Grace Bio-Labs, Inc.