Principaux documents
ETIPRP20-EP
Eppendorf® epT.I.P.S. Reloads, PCR Clean
volume × L 0.5-20 μL × 46 mm, (light gray), pack of 960 ea (10 trays × 96 tips)
960 EA
187,00 $
187,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
960 EA
187,00 $
About This Item
Code UNSPSC :
41121600
187,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
(light gray)
Taille
L
Stérilité
non-sterile
Caractéristiques
DNase free
RNase free
PCR Clean
Conditionnement
pack of 960 ea (10 trays × 96 tips)
Fabricant/nom de marque
Eppendorf® 0030073789 (EU)
Eppendorf® 022491725 (NA)
Technique(s)
PCR: suitable
Volume maximum
20 μL
Volume × L
0.5-20 μL × 46 mm
Volume
0.5—20 μL
Activité étrangère
PCR inhibitors, Human DNA, DNase, Rnase, free
Catégories apparentées
Description générale
Part of the ep T.I.P.S. box and reload system for contamination-free handling of refill trays, with plastic lids protecting the trays from contact during transfer to the working box.
Trays of tips which may be autoclaved in their packaging and stored clean for later use. Tips may be used straight from the reload tray, or loaded into ep T.I.P.S box (purchase separately). PCR clean, free of human DNA, DNase, RNase and PCR inhibitors, batch related certificates available on the internet at http://www.eppendorf.com
Informations légales
Eppendorf is a registered trademark of Eppendorf AG