Principaux documents
EPM1344-0009
Eppendorf® New Brunswick I24/I24R Incubator Shaker
I24R, refrigerated, orbit diameter 3/4 in., AC/DC input 120 V, 50 - 60 Hz, US 3-pin plug
About This Item
Code UNSPSC :
41104405
Caractéristiques
CSA and CE compliant
I24R
orbit diameter 3/4 in.
refrigerated
Fabricant/nom de marque
Eppendorf® M1344-0009 (US)
Paramètres
-15-60 °C temp. range
25-500 rpm (speed range)
Larg. × P × H
56 cm × 76.2 cm × 61 cm
Taille de la plateforme
46 cm × 46 cm
Poids
60 kg
Description générale
benchtop incubator with built-in shaker
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
New Brunswick is a trademark of Eppendorf AG