EPM1231-9934
Platform for Eppendorf® New Brunswick™ Innova® 3100 Water Bath Shaker
for 125 mL Erlenmeyer flasks, dedicated, interchangeable
1 600,00 $
About This Item
1 600,00 $
Matériaux
stainless steel
Caractéristiques
dedicated
for 125 mL Erlenmeyer flasks
interchangeable
Fabricant/nom de marque
Eppendorf® M1231-9934 (NA)
L × l
30.5 cm × 42 cm