Principaux documents
EP6731010025
Eppendorf® CellXpert® C170i CO2 Incubator
Std, Left, AC/DC input 120 V, US 2-pin plug
Synonyme(s) :
Eppendorf CO2 Incubator
1 EA
15 440,00 $
15 440,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
15 440,00 $
About This Item
Code UNSPSC :
41102404
15 440,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
Std, Left
Fabricant/nom de marque
6731010025
Eppendorf®
Paramètres
4-50 °C temp. range
Capacité
(170 L)
(740 L)
Catégories apparentées
Description générale
Stay flexible for the future
  • Upgrade your device later in your lab (e.g. door handle position)
  • Advanced user interface for easy and complete control over parameter settings, monitoring, and documentation
Safety for your cells
  • Uniform temperature with multiple sensors verified at 27 points (German Norm DIN 12880)
  • Fast temperature and CO2 recovery in less than 5 minutes without overshoot
  • Vibration and turbulence protection with fan-less design
  • 180 ºC High Temperature Disinfection (HTD)
Save money
  • Significantly reduced gas consumption with smart gas control
  • Up to 25 % more usable space with fan-less design
  • No recurring costs for internal replacement parts (HEPA filters, UV lamps etc.)
Informations légales
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG