Principaux documents
EP5804760000
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds 50-75 mL tubes, pack of 2 ea
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Caractéristiques
holds 50-75 mL tubes
Conditionnement
pack of 2 ea
Fabricant/nom de marque
Eppendorf® 5804760000
Technique(s)
centrifugation: suitable
Compatibilité
for use with 100 mL rectangular bucket
Description générale
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG