Principaux documents
EP4960800090
Eppendorf® Bottle Thread Adapter
from GL 32 to NS 24/29, polypropylene
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.17
Matériaux
polypropylene
Caractéristiques
from GL 32 to NS 24/29
Fabricant/nom de marque
Eppendorf® 4960800090
Description générale
Eppendorf dispensers are designed for bottle threads of 45 mm. For other bottle threads a thread adapter can be used. The diameter of the thread adapter is imprinted on the adapter. If the bottle thread is smaller than 32 mm, a second adapter must be used. If you need a thread adapter with a higer resistance to chemicals, use a PTFE/ETFE adapter.
Application
Connection of Eppendorf dispensers to bottles with neck threads different to 45 mm.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG