Principaux documents
EP4861000834
Eppendorf® Xplorer® Move It pipette
no. of channels, 6, cone volume × distance 50-1,200 μL × 9 mm, green
1 EACH
4 130,00 $
4 130,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EACH
4 130,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.01
4 130,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
green
Caractéristiques
no. of channels 6
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 4861000834
Volume du cône × distance
50-1,200 μL × 9 mm
Description générale
  • Adjustable tip spacing pipette
  • Multi-channel pipette
  • Electronic pipette
  • 6-channel
  • 50-1,200μL
  • 9mm cone distance
  • Green
Application
  • Automated pipetting of liquids (Pip) for forward pipetting
  • Manual pipetting (Man)for volume determination
  • Pipetting and mixing (P/M) for automated mixing of samples
  • Dispensing (Dis)for manual filling of plates, gels, and reaction vessels
  • Automatic dispensing (Ads for automated filling of plates, gels, and reaction vessels
  • Multi aspiration: Pooling supernatants from different vessels (SPC)
  • Sequential dispensing and pipetting (Spc) for user-defined volume steps
  • Reverse pipetting (Spc) for non-aqueous solutions
  • Diluting (Spc) for preparation of dilution series
  • Fix volume (Fix): Save up to 10 volumes for ease of operation
Caractéristiques et avantages
Air cushioned pipettes with variable volume and adjustable tip spacing
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Xplorer is a registered trademark of Eppendorf AG