Key Documents
EP022822225
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-45-8-17, non aerosol tight, (8 x 15 mL conical tubes)
About This Item
Code UNSPSC :
41103909
Caractéristiques
to hold 8 x 15 mL conical tubes
to hold 8 x 17 x 118 - 123 mm tubes
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022822225 (NA)
Technique(s)
centrifugation: suitable
Capacité
(8 x 15 mL conical tubes)
Compatibilité
for use with centrifuge models Vacufuge Plus Concentrator
Catégories apparentées
Informations légales
Eppendorf is a registered trademark of Eppendorf AG
Vacufuge is a registered trademark of Eppendorf AG