EP022822179
Rotor for Eppendorf® Vacufuge® Plus Concentrator
F-50-8-18, non aerosol tight, (8 x 10 mL - 18 mL round-bottom tubes (18 x 105-128 mm))
1 460,00 $
About This Item
1 460,00 $
Caractéristiques
to hold 8 x 10/18 mL round-bottom tubes
to hold 8 x 18 x 105 - 128 mm tubes
fixed angle rotor
non aerosol tight
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Eppendorf® 022822179 (NA)
Technique(s)
centrifugation: suitable
Capacité
(8 x 10 mL - 18 mL round-bottom tubes (18 x 105-128 mm))
Compatibilité
for use with centrifuge models Vacufuge Plus Concentrator