Documents
EP0030609301
Eppendorf® Microplate 96/V-PP, 96 wells
def. color (wells colorless), PCR clean, with barcode, polypropylene, pkg of 80 ea
About This Item
Code UNSPSC :
41122107
Matériaux
polypropylene
Conditionnement
pkg of 80 ea
Fabricant/nom de marque
Eppendorf® 0030609301
Technique(s)
PCR: suitable
Taille
96 wells
Informations légales
Eppendorf is a registered trademark of Eppendorf AG