Principaux documents
EP0030128486
Eppendorf® Twin tec PCR plates, 384 wells
with barcode, individual color (wells colorless), pkg of 25 ea
About This Item
Code UNSPSC :
41106300
Conditionnement
pkg of 25 ea
Fabricant/nom de marque
Eppendorf® 0030128486
Technique(s)
PCR: suitable
Taille
384 wells
Description générale
The Eppendorf twin.tec PCR plate combines extremely thin-walled polypropylene wells for optimal heat transfer to the sample, with robust polycarbonate frames for high rigidity. With raised well rims, twin.tec plates enable highly effective sealing to reduce the risk of cross-contamination. All Eppendorf twin.tec PCR plates are ‘PCR clean’, having been batch-tested and certified by an independent laboratory to be free of a range of common PCR contaminants, including DNA, DNAse, RNAse, and PCR inhibitors. With twin.tec PCR plates you’ll have maximum confidence in your results.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG