EP0030127749
Eppendorf® Serological Pipets, Sterile Grade
violet, capacity 50 mL, pack of 160 ea (4 bags × 40 pcs.)
552,00 $
About This Item
552,00 $
Produits recommandés
Matériaux
violet
Stérilité
sterile
Caractéristiques
DNA free
graduations
plugged
Conditionnement
pack of 160 ea (4 bags × 40 pcs.)
Fabricant/nom de marque
Eppendorf® 0030127749 (Global)
Capacité
50 mL
Description générale
- Eppendorf® Serological Pipets are manufactured from ultra-pure virgin polystyrene for optimal clarity.
- Easy-to-read graduations for accurate volume determination.
- Pipets are color-coded based on sizes for easy identification.
- Sterility assurance level of 10-6.
- Compatible with all mechanical and electronic pipette controllers - including Pipet Helper® or Easypet® 3.
- Free of detectable pyrogens, DNA, RNase and DNase and certified non-cytotoxic.
Application
- Transfer of liquids
- Mixing suspensions
- Combining reagents and chemical solutions
- Transferring cells for empirical analysis or expansion
- Layering reagents for creating higher density gradients