Principaux documents
EP0030089545
Eppendorf® Combitips Advanced
volume 1.0 mL, yellow, sterile, pack of 100 ea (individually wrapped)
Synonyme(s) :
pipetting tips
100 EACH
266,00 $
266,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EACH
266,00 $
About This Item
Code UNSPSC :
41121600
266,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
polypropylene
yellow
Stérilité
sterile
Caractéristiques
graduation: no
Conditionnement
pack of 100 ea (individually wrapped)
Fabricant/nom de marque
Eppendorf® 0030089545
Volume
1.0 mL
Catégories apparentées
Description générale
Eppendorf Combitips are our tip of choice when working with Multipette multiple dispensing pipettes. Combitips advanced dispenser tips offer you many advantages for working in a modern laboratory thanks to their revolutionary design. With an integrated piston, they work according to the positive displacement principle and ensure that the correct volume of liquid is dispensed regardless of the physical properties of the liquid you are working with (e.g. viscosity, volatility, density, temperature). This allows you to dispense with high precision regardless of the flow properties of the liquid and, thanks to the hermetically sealed piston that prevents contamination by aerosols, you can work safely with radioactive or toxic materials. Eppendorf pioneered the Multipette/Combitips multiple dispensing system in 1978 and has improved it several times since then. This was the very first pipetting system that allowed the user to fill the tip once and dispense multiple times. The concept continues to be a driver for efficient liquid handling and is the tool of choice in many laboratories working with plates or series of reaction vessels. In 2023, the Combitips advanced pipette tips were certified with ACT Environmental Impact Factor labels by the non-profit organization My Green Lab . This makes them the ideal choice for laboratories looking for more sustainable products.
Caractéristiques et avantages
  • Direct displacement principle (similar to a syringe)
  • High-precision dispensing regardless of the physical properties of the liquid (e.g. viscosity, volatility, density or temperature)
  • Prevents aerosol contamination through the hermetically sealed piston and thus ensures safe dispensing
  • Protection against radioactive and toxic substances
  • Fast dispensing of long series through precise, repeated dosing of identical volumes (in combination with Multipette hand dispensers)
Informations légales
Eppendorf is a registered trademark of Eppendorf AG