EP0030078853
Eppendorf® ep Dualfilter T.I.P.S.
PCR clean, sterile, working volume 0.1-20 μL, L 42 mm, rose, pack of 3840 ea (10 racks of 384 tips)
Synonyme(s) :
Eppendorf® ep Dualfilter T.I.P.S.®, pipette tip
1 180,00 $
About This Item
1 180,00 $
Produits recommandés
Matériaux
rose
Stérilité
sterile
Caractéristiques
PCR clean
Conditionnement
pack of 3840 ea (10 racks of 384 tips)
Fabricant/nom de marque
Eppendorf® 0030078853
Longueur
42 mm
Volume
0.1—20 μL
Volume de travail
0.1-20 μL
Catégories apparentées
Description générale
Caractéristiques et avantages
- Two filter layers provide dual protection for pipette and sample
- Two filter layers provide dual protection from aerosols and biomolocules
- Free from PCR inhibitor additives
- Patent pending technology
- All tips are provided sterile, Eppendorf PCR clean and pyrogen-free
- Defined flow dynamicsLow wettability
- High thermal stability
- Leak proof seal
- Minimum attachment and ejection forces