Principaux documents
EP0030077539
Eppendorf® ep Dualfilter T.I.P.S.®
10 racks x 96 ea, PCR clean, capacity 2-20 μL, sterile
About This Item
Code UNSPSC :
41121600
Le produit EP0030077539 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Matériaux
yellow (Tray)
Stérilité
sterile
Caractéristiques
DNase free
RNase free
filter
Conditionnement
pack of 960 ea (10 racks of 96 tips)
Fabricant/nom de marque
Eppendorf® 0030077539
Technique(s)
PCR: suitable
Capacité
2-20 μL
Volume maximum
20 μL
Volume
2—20 μL
Description générale
  • Eppendorf® ep Dualfilter T.I.P.S.® (2-20 μL) are uncoated, free of additives and do not leach into the sample.
  • Ultrahomogenous surface for maximum reproducibility.
  • Ultrahydrophobic surface for minimal sample loss.
  • Significantly reduced foam formation during pipetting.
  • Extreme resistance to chemicals.
  • Available in PCR clean and Eppendorf Quality.
  • Also available as ep Dualfilter T.I.P.S. in PCR clean/Sterile
  • Sterile and pyrogen-free.
  • No Coating.
Application
  • Cell culture (media).
  • Genomics: PCR, RT-PCR, qPCR and all other types of PCR.
  • Enzymatic reactions (restriction decomposition, ligation).
  • Isolation and purification of nucleic acid.
  • Gel electrophoresis (e.g., prefabricated DNA ladders) typical detergents: SDS, Triton, X-100, Brij® 35, Tween® 20, CHAPS.
  • Proteomics (all types of protein examinations).
  • Protein isolation and purification.
  • NGS library preparation.
Informations légales
Brij is a registered trademark of Croda International PLC
Eppendorf is a registered trademark of Eppendorf AG
TWEEN is a registered trademark of Croda International PLC
Triton is a trademark of The Dow Chemical Company or an affiliated company of Dow
ep Dualfilter T.I.P.S. is a registered trademark of Eppendorf AG