DWP96500G4-EP
Eppendorf® Deepwell Plate 96/500μL
PCR Clean, volume 500 μL, green border with clear wells, pkg of 40 plates (5 bags x 8 plates)
522,00 $
About This Item
522,00 $
Produits recommandés
Stérilité
non-sterile
Caractéristiques
PCR Clean
green border with clear wells
Conditionnement
pkg of 40 plates (5 bags x 8 plates)
Fabricant/nom de marque
Eppendorf® 951031861
Disponibilité
available only in North America
Technique(s)
PCR: suitable
Taille
96 well
Volume
500 μL
Puits
96
Catégories apparentées
Description générale
Caractéristiques et avantages
- OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors via high-contrast alphanumeric labeling
- RecoverMax® well design: Optimized well geometry for maximum sample recovery and excellent mixing properties
- Minimal remaining volume and high uniformity from well to well
- Raised well rims and smooth surface for reliable closing, even with heat sealing
- Easy and reliable stacking
- g-Safe: exceptional centrifugation stability up to 6,000 × g for faster protocols and improved sample quality
- High-quality polypropylene guarantees high resistance to chemicals, mechanical stress and temperature extremes
- Manufactured without slip agents, plasticizers, and biocides – Materials which could have a negative effect on bioassays
- Autoclavable (121?°C, 20?min)
- Available with barcode. Additional details available at: www.eppendorf.com/barcode