Principaux documents
CLS6705
Corning® LSE Platform Rocker
Double PlatformDouble Platform, AC/DC input 115 V, AC/DC input 50/60 Hz, AC/DC input, US 3-pin plug
Synonyme(s) :
Laboratory rocker, Platform shaker, Rocking platform
1 EA
1 620,00 $
1 620,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 620,00 $
About This Item
Code UNSPSC :
41103811
1 620,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
Double Platform
Double Platform
Fabricant/nom de marque
Corning 6705
6705
Description générale
The Corning LSE Platform Rocker is a compact yet high capacity rocker that is ideal for western blotting and general mixing applications. The adjustable rocking speed and fixed 7 degree tilt makes it optimal for staining fragile gels, as well as the more intensive action necessary for washing blots.
The speed of the Corning Platform Rocker can be adjusted from 2 to 60 rpm and the timer can be set for up to 2 hours or for continuous operation. The unit is safe to use in temperature controlled environments up to 70°C and is available in single or high capacity double platform configurations.
Features and Benefits
  • Low profile and compact size
  • 11-pound load capacity
  • Non-slip rubber pads to prevent sample containers from moving during operation
  • Double platform model available for increased capacity
Informations légales
Corning is a registered trademark of Corning, Inc.
LSE is a trademark of Corning, Inc.