Principaux documents
CLS432077
Corning® CoolCell Filler Vials, for use with CoolCell LX and CoolCell FTS30 containers
5 mL
Synonyme(s) :
centrifuge tubes
6 EA
43,80 $
43,80 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
6 EA
43,80 $
About This Item
Code UNSPSC :
41103025
Nomenclature NACRES :
NB.12
43,80 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pkg of 6
Fabricant/nom de marque
Corning 432077
Capacité
5 mL
Description générale
To ensure cell freezing rate consistency and uniform results when using CoolCell containers, insert a CoolCell Filler Vial into empty wells when freezing less than a full load. It is suitable for repeated use and compatible with CoolCell LX, CoolCell FTS30 and CoolCell 5ml LX containers.
Informations légales
CoolCell is a trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.