Principaux documents
CLS3809
Corning® 96 Well CellBIND® Microplates
white polystyrene plate, sterile, flat bottom clear wells, lid, case of 100 plates
Synonyme(s) :
cell culture plate, multi well plates, multiple well plates
100 EA
889,00 $
889,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
100 EA
889,00 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.13
889,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
flat bottom clear wells
flat bottom wells
polystyrene plate
white polystyrene plate
Stérilité
sterile
sterile
Caractéristiques
lid
lid
plate format: 96 well standard
Conditionnement
case of 100 plates
pack of 5
case of 50
Fabricant/nom de marque
Corning 3809
Taille
96 wells
Superficie
0.32 cm2 , cell growth area
cell growth area
cell growth area
Volume des puits
360 μL
Volume de travail par puits
75-200 μL
Adéquation
suitable for (cell cuture)
Description générale
  • Corning® 96 Well CellBIND® Microplates have an optically clear flat bottom for direct microscopic viewing.
  • Thinner bottom enables lower background fluorescence and allows readings down to 340 nm.
  • Opaque walls aid in minimal well-to-well cross-talk.
  • Flat bottoms with 360 μL total volume
  • Recommended working volumes of 75 to 200 μL
  • CellBIND® surface treated for optimal cell attachment
  • Nonreversable lids with condensation rings to reduce contamination
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Individual alphanumeric codes for well identification
Informations légales
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.