CLS3121STPK
Replacement stopcock, for Pyrex® small top desiccators with 24/29 standard taper stopcock
replacement 24/29 stopcock w/ hose connector, 1/cs
Synonyme(s) :
Corning stopcock
157,00 $
About This Item
157,00 $
Produits recommandés
Matériaux
borosilicate glass (borosilicate glass code 7740)
Joint
joint ST/NS24/29
Caractéristiques
reusable
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
Corning 3121-STPK
Hauteur
117 mm