Principaux documents
MS
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
About This Item
Code UNSPSC :
41106514
Source biologique
African green monkey kidney
Conditionnement
tube of 5 μg 91070510-DNA-5UG
pkg of vial of cells 91070510-1VL
Mode de croissance
Adherent
Caryotype
Not specified
Morphologie
Epithelial
Récepteurs
Not specified
Maladie(s) pertinente(s)
cancer
Origine de la lignée cellulaire
Monkey African Green kidney
Description de la lignée cellulaire
Derived from African green monkey. Useful for supporting functional co-cultures of rat hepatocytes. Originally mycoplasma infected.
Milieu de culture
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 5-10% FBS / FCS (F2442).
Procédure de repiquage
Split sub-confluent cultures (70-80%) 1:3 to 1:6, i.e., seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.
Autres remarques
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.