Principaux documents
AXYVT38431ULRS
Pipette tips in 384-tip format, robotic
size 30 μL, pkg of 5x3840tips/cs, racked, sterile
Synonyme(s) :
Pipette tips for Velocity
1 PKG
5 510,00 $
5 510,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
5 510,00 $
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
5 510,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Stérilité
sterile
Fabricant/nom de marque
Axygen Inc VT-384-31UL-R-S
Corning
Conditionnement
pkg of 5x3840tips/cs
Taille
30 μL
Volume
30 μL
Description générale
  • Axygen® automation pipette tips in 384-tip format are designed for Agilent®/Velocity11 VPrep® and Bravo 96LT, 8LT, 384ST, 96ST, and 16ST head.
  • Manufactured to stringent specifications including automated assembly to minimize contamination risks and ensure consistent performance and quality.
  • The clear polypropylene high-quality tips are ideal for sensitive biological applications.
  • RNase-/DNase-free and nonpyrogenic.
Autres remarques
Browse all pipette tips
Informations légales
Agilent is a registered trademark of Agilent Technologies, Inc.
Axygen is a registered trademark of Corning, Inc.
Velocity11 VPrep is a registered trademark of Corning, Inc.