Documents
AXYPCR96CS
Corning® Axygen® PCR Microplate
size 96 wells, sterile, conical bottom clear polypropylene wells
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
conical bottom clear polypropylene wells
Stérilité
sterile
Fabricant/nom de marque
Axygen Inc PCR-96-C-S
Corning
Volume maximum
200 μL
Taille
96 wells
Volume de travail
200 μL
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.