Principaux documents
AXYPCR384Y
Corning® Axygen® PCR Microplate
size 384 wells, conical bottom yellow polypropylene wells, pkg of 5x10plates/cs
Synonyme(s) :
384 multiwell plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
conical bottom yellow polypropylene wells
Stérilité
non-sterile
Fabricant/nom de marque
Axygen Inc PCR-384-Y
Corning
Conditionnement
pkg of 5x10plates/cs
Volume maximum
40 μL
Taille
384 wells
Volume de travail
40 μL
Adéquation
suitable for PCR
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.