Principaux documents
AXYPCR38455C
Corning® Axygen® PCR Microplate
size 384 wells, conical bottom black polypropylene wells, maximum volume 55 μL, pkg of 5x10plates/cs
Synonyme(s) :
384 multiwell plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
conical bottom black polypropylene wells
Stérilité
non-sterile
Fabricant/nom de marque
Axygen Inc PCR-384-55-C
Corning
Conditionnement
pkg of 5x10plates/cs
Volume maximum
55 μL
Taille
384 wells
Volume de travail
50 μL
Adéquation
suitable for (thermal cycling)
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.