Principaux documents
SHORT09NC
BS/IP/SL(S) Viscometer for Transparent Liquids
Size 9, Nom. Constant 10, Range 2,000 to 10,000 cSt, (non certified)
About This Item
Code UNSPSC :
41116107
Nomenclature NACRES :
NA.24
Fabricant/nom de marque
Paragon Scientific Ltd
Application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Description générale
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product SHORT09UKC