Principaux documents
GX308070528
Supelco
Quartz Torch
High Solids Full Length with 90 Deg. Bend & 2.3 mm Injector
About This Item
Code UNSPSC :
12352200
Nomenclature NACRES :
SB.32
Qualité
High Solids Full Length with 90 Deg. Bend & 2.3 mm Injector
for 700-ES or Vista Axial
Fabricant/nom de marque
(30-807-0528)
Compatibilité
Agilent/Varian 700-ES Series Axial, Vista Axial