Principaux documents
GX044GYF
Supelco
Contour Flared End PVC Pump Tube
2tag 0.44mm ID Green/Yellow (PKT 6)
About This Item
Code UNSPSC :
12352200
Nomenclature NACRES :
SB.32
Fabricant/nom de marque
0.44-GY-F
Compatibilité
Spectro/Ametek EOP (Modula, CIROS, Genesis, ARCOS)
Spectro/Ametek SOP (Modula, CIROS, Genesis, ARCOS)
Spectro/Ametek Spectro Blue EOP
Spectro/Ametek Spectro Blue SOP