Principaux documents
SPKGENELC045
Millipore
Spare part, filter mat
W × H × D 173 mm × 173 mm × 17 mm
About This Item
Code UNSPSC :
23151806
Matériaux
fibrous
Niveau de qualité
Gamme de produits
Mobius®
Larg. × H × P
173 mm × 173 mm × 17 mm
Compatibilité
for use with Mobius®
Description générale
These filters are used on BioProcessing systems to capture dust, dirt, and other airborne particles. They ensure optimal performance and of the equipment.
This spare part is required for the preventive maintenance service of some of our BioProcessing systems, it is recommended to have one in stock and to change it every 12 months.
Conditionnement
Pack of 5 units
Informations légales
MOBIUS is a registered trademark of Merck KGaA, Darmstadt, Germany