Principaux documents
Z768065
Flask carriers for RCT and RET magnetic stirrers
fits flask, 2000 mL without handles
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.72
Caractéristiques
fits flask 2000 mL without handles
Conditionnement
pack of 1 ea
Fabricant/nom de marque
IKA 0025000834
Description générale
Flask carrier without handle for use IKA stirrers, can hold 1 x 2000ml round bottom flask. Outside diameter 194.7 mm