Principaux documents
Z755796
Block for MultiTherm shaker
96x 0.2 mL or 1 x PCR plate
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NB.76
Caractéristiques
Holds 1 x plate
Holds 96 X 0.2 mL tubes
Fabricant/nom de marque
Benchmark Scientific H5000-02
Technique(s)
PCR: suitable
Description générale
An assortment of blocks are available to use with the MultiTherm shaker, each sold separately.
Informations légales
MultiTherm is a trademark of Benchmark Scientific, Inc.