Principaux documents
Z743293
Julabo® adaptors
quad distributing, 1 1/4 inch female with barbed fittings, for 1 inch i.d. tubing, pack of 2
Synonyme(s) :
barbed adaptor
2 EA
4 220,00 $
4 220,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
2 EA
4 220,00 $
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
4 220,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pack of 2
Fabricant/nom de marque
Julabo 8970524
Description générale
For use with Julabo® FL Series Recirculating Cooler / Chillers , 2 Quad distributing adapters, G 1 1/4″ female, with barbed fittings for tubing 1″ ID. Set of two circular quad manifolds with internal baffles for constant water pressure and temperature to all outlets. G 1¼” female thread fittings (which can be fitted with shut off valves), comes complete with all barbed fittings for 1” ID tubing.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.