Principaux documents
Z743272
Julabo® adaptors
barbed fittings, for 10 mm i.d. tubing, pack of 2
Synonyme(s) :
barbed adaptor
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Conditionnement
pack of 2
Fabricant/nom de marque
Julabo 8970447
Description générale
For use with Julabo® circulators, 2 barbed fittings for tubing 10 mm ID. For use with M16x1 fittings. Requires a female M16x1 cap nut.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.