Principaux documents
Z743202
Julabo® bath carrier trays for SW22/SW23
spring clamps for use with universal basic tray
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Fabricant/nom de marque
Julabo 8970631
Description générale
Spring clamps set for use with basic stainless steel tray Z743203 (8970630). Set includes 12 spring clamps 135 mm long and 5 spring clamps 190 mm long. For use with Julabo® shaking water bath models SW22, SW23.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.