Z742839
Kern PCB series precision balances, rechargable battery pack
pack of 1 ea
About This Item
Code UNSPSC :
41111502
Nomenclature NACRES :
NB.71
Produits recommandés
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Kern PCB-A01
Description générale
Internal rechargeable NiMH battery pack PCB-A01, with Euro, UK, US, CH plug types. Operating time up to 48 h, charging time approx. 8 h, Auto-Off function preserves the rechargeable battery pack. Suitable for models from the KERN PCB, KERN ECB, KERN ECE and KERN FKB-A series.