Principaux documents
Z692867
BRAND® HandyStep® S manual repetitive pipette
Synonyme(s) :
Adjustable volume pipette
1 EA
664,00 $
664,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
664,00 $
About This Item
Code UNSPSC :
41121500
Nomenclature NACRES :
NB.02
664,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
electronic: no
single-channel
Conditionnement
pack of 1 ea
Fabricant/nom de marque
BRAND 705110
Plage de mesure du volume
2 μL-5 mL
Volume
5 mL
Description générale
Ideal for routine or research applications in diagnostics, molecular biology, environmental analysis, and many more.
HandyStep S and PD-tips work on the direct displacement principle. This offers the highest-precision dispensing of liquid media with high viscosity, high density, or high vapor pressure. Direct displacement permits contamination-free operation, since no aerosols are formed.
  • Excellent chemical resistance through innovative plastic materials
  • Easy tip mounting - PD-tip is now simply inserted from below
  • Dispensed volume range from 2 μL to 5 mL
  • Up to 49 dispensing steps
  • Lightweight and easy to handle
  • Suitable for use with BRAND PD-tips, Encode tips, Repet tips, Combitips®
Includes:
HandyStep S, conformity-certified, with serial number, performance cerficate, operating manual, shelf/rack mount, 3 PD-Tips: 0.1 mL, 1 mL and 10 mL.
Caractéristiques et avantages
  • Serial pipetting made easy
  • Works with BRAND PD-Tips and third-party tips
  • Robust, stable and durable
Autres remarques
BR705110 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
Combitips is a registered trademark of Eppendorf AG
HandyStep is a registered trademark of BRAND GMBH + CO KG