Principaux documents
Z680443
Duran® GL 45 connection system caps and accessories
hose connection screw cap, blue PP GL 14
Synonyme(s) :
1129814
2 EA
32,90 $
32,90 $
Disponible pour expédition le22 avril 2025Détails
Changer de vue
2 EA
32,90 $
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
32,90 $
Disponible pour expédition le22 avril 2025Détails
Matériaux
blue polypropylene
Fabricant/nom de marque
DURAN® 1129814
Paramètres
-40-140 °C temp. limit
Description générale
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Caractéristiques et avantages
  • Manufactured from PP, PTFE, silicone
  • Accessories designed for use with the DURAN® GL 45 2/3 port tubing connection caps or GL 45 / GLS 80® Stirred Bottle Reactor caps
  • Safe and secure liquid transfer into or out of laboratory bottles
  • Four inserts are suitable for rigid or semi-rigid tubing with a range of outer diameters: 1.6, 3.0, 3.2 or 6.0 mm
  • Open-topped blue PP GL 14 pass-through tubing cap for use with the inserts
  • Closed-top red PBT GL 14 cap for sealing unused ports
  • Sterile pressure equilisation is possible with pressure compensation set
Informations légales
DURAN is a registered trademark of DWK Life Sciences
GLS 80 is a registered trademark of Duran Group
Questions
  1. Does this item include all the pieces shown in the photo, or is that a different SKU?
    1 answer
    1. The photo provided is representative of the entire group of related GL 45 capping products. Each component is offered separately. This specific product is the blue hose connection screw cap shown second from the right as seen in provided image.